Assay Details
Target Gene Details
Entrez Gene ID: | 3184 |
Gene Name: | heterogeneous nuclear ribonucleoprotein D |
Gene Aliases: |
AUF1, AUF1A, HNRPD, P37, hnRNPD0 |
Location: |
Chr.4:82353314-82373996 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPD | NM_001003810.1 | 7 | 1651 | NP_001003810.1 |
| NM_002138.3 | 8 | 1708 | NP_002129.2 | |
| NM_031369.2 | 8 | 1798 | NP_112737.1 | |
| NM_031370.2 | 9 | 1855 | NP_112738.1 | |
| AF039575.1 | 8 | 1366 | AAB96683.1 | |
| BG180865.1 | 1 | 193 | ||
| BG180941.1 | 1 | 194 | ||
| BM832779.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv594743 | Chr.4:82353481 - 82404421 on Build GRCh38 | Loss |
|
| nsv829988 | Chr.4:82316984 - 82489338 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map