Assay Details
Target Gene Details
Entrez Gene ID: | 5934 |
Gene Name: | RB transcriptional corepressor like 2 |
Gene Aliases: |
P130, Rb2 |
Location: |
Chr.16:53434420-53491648 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBL2 | NM_001323608.1 | 9 | 1389 | NP_001310537.1 |
| NM_001323609.1 | 9 | 1389 | NP_001310538.1 | |
| NM_001323610.1 | 9 | 1389 | NP_001310539.1 | |
| NM_001323611.1 | 9 | 1138 | NP_001310540.1 | |
| NM_005611.3 | 9 | 1370 | NP_005602.3 | |
| XM_011523253.2 | 7 | 763 | XP_011521555.1 | |
| XM_017023513.1 | 6 | 927 | XP_016879002.1 | |
| AK300047.1 | 9 | 1108 | ||
| AK304283.1 | 6 | 837 | ||
| AK307881.1 | 9 | 1347 | ||
| BC034490.1 | 9 | 1360 | AAH34490.1 | |
| BX537767.1 | 8 | 951 | CAD97830.1 | |
| S67171.1 | 9 | 1280 | AAB29227.1 | |
| X74594.1 | 9 | 1081 | CAA52671.1 | |
| X76061.1 | 9 | 1321 | CAA53661.1 |
More Information
Set Membership: |
|

Genomic Map