Assay Details
Target Gene Details
Entrez Gene ID: | 200810 |
Gene Name: | ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase like |
Gene Aliases: |
ALG1L1 |
Location: |
Chr.3:125929272-125993832 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALG1L | XM_017005844.1 | XP_016861333.1 | ||
| XM_017005846.1 | XP_016861335.1 |
Target Gene Details
Entrez Gene ID: | 152015 |
Gene Name: | rhophilin associated tail protein 1B |
Gene Aliases: |
- |
Location: |
Chr.3:125969144-125983454 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ROPN1B | NM_001012337.2 | NP_001012337.1 | ||
| NM_001308313.1 | NP_001295242.1 | |||
| XM_005247138.3 | XP_005247195.1 | |||
| XM_006713513.3 | XP_006713576.1 | |||
| AF231410.1 | AAG59586.1 | |||
| BC015413.1 | AAH15413.1 | |||
| BC141849.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591465 | Chr.3:125283331 - 126140524 on Build GRCh38 | Gain |
|
| nsv829716 | Chr.3:125928159 - 126116976 on Build GRCh38 | Gain+Loss |
|
| nsv1011457 | Chr.3:125287676 - 126146923 on Build GRCh38 | Gain |
|
| nsv3984 | Chr.3:125946515 - 125993155 on Build GRCh38 | Deletion |
|
| nsv967059 | Chr.3:125961987 - 125987093 on Build GRCh38 | Duplication |
|
| esv2759174 | Chr.3:125822490 - 125983913 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map