Assay Details
Target Gene Details
Entrez Gene ID: | 79018 |
Gene Name: | GID complex subunit 4 homolog |
Gene Aliases: |
C17orf39, VID24 |
Location: |
Chr.17:18039297-18068405 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GID4 | NM_024052.4 | 6 | 2385 | NP_076957.3 |
| XM_005256800.1 | 5 | 2141 | XP_005256857.1 | |
| AK124377.1 | 1 | 501 | ||
| AK127580.1 | 6 | 2072 |

Genomic Map