Assay Details
Target Gene Details
Entrez Gene ID: | 23451 |
Gene Name: | splicing factor 3b subunit 1 |
Gene Aliases: |
Hsh155, MDS, PRP10, PRPF10, SAP155, SF3b155 |
Location: |
Chr.2:197391974-197435093 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SF3B1 | NM_012433.3 | 25 | 3987 | NP_036565.2 |
| AF054284.1 | 25 | 3893 | AAC97189.1 | |
| AF070540.1 | 5 | 865 | AAC28633.1 | |
| AL137410.1 | 3 | 410 | CAB70728.1 | |
| CD692669.1 |
More Information
Set Membership: |
|

Genomic Map