Assay Details
Target Gene Details
Entrez Gene ID: | 79707 |
Gene Name: | nucleolar protein 9 |
Gene Aliases: |
Grc3, NET6 |
Location: |
Chr.1:6521347-6554598 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOL9 | NM_024654.4 | 12 | 2536 | NP_078930.3 |
| XM_005263493.3 | 10 | 1868 | XP_005263550.1 | |
| XM_011542147.2 | 7 | 1462 | XP_011540449.1 | |
| XM_017002338.1 | 10 | 2046 | XP_016857827.1 | |
| XM_017002339.1 | 9 | 1762 | XP_016857828.1 | |
| AK026976.1 | 8 | 1546 | BAB15611.1 | |
| AK091284.1 | 12 | 2473 | ||
| BC009257.2 | AAH09257.2 | |||
| BF969522.1 | ||||
| BU728251.1 | 1 | 380 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7880 | Chr.1:6417968 - 6611914 on Build GRCh38 | Gain+Loss |
|
| nsv545268 | Chr.1:6490316 - 6578004 on Build GRCh38 | Loss |
|
| nsv831536 | Chr.1:6429326 - 6605343 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map