Assay Details
Target Gene Details
Entrez Gene ID: | 5884 |
Gene Name: | RAD17 checkpoint clamp loader component |
Gene Aliases: |
CCYC, HRAD17, R24L, RAD17SP, RAD24 |
Location: |
Chr.5:69369297-69414801 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD17 | NM_001278622.1 | NP_001265551.1 | ||
| NM_002873.1 | NP_002864.1 | |||
| NM_133338.2 | NP_579916.1 | |||
| NM_133339.2 | NP_579917.1 | |||
| NM_133340.2 | NP_579918.1 | |||
| NM_133341.2 | NP_579919.1 | |||
| NM_133342.2 | NP_579920.1 | |||
| NM_133343.1 | NP_579921.1 | |||
| NM_133344.2 | NP_579922.1 | |||
| XM_017009679.1 | XP_016865168.1 | |||
| XM_017009680.1 | XP_016865169.1 | |||
| XM_017009681.1 | XP_016865170.1 | |||
| AF017748.1 | AAD01620.1 | |||
| AF076838.1 | AAC95520.1 | |||
| AF085736.2 | AAC36334.1 | |||
| AF098533.1 | AAC97950.1 | |||
| AF098534.1 | AAC97951.1 | |||
| AF112263.1 | AAD38878.1 | |||
| AF126424.1 | AAD17334.1 | |||
| AJ001642.1 | CAA04894.1 | |||
| AJ004977.1 | CAA06251.1 | |||
| AK292487.1 | ||||
| AL122068.1 | CAB59244.1 | |||
| BC032304.1 | AAH32304.1 | |||
| BX537441.1 | CAD97683.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5694n100 | Chr.5:69081291 - 69374274 on Build GRCh38 | Gain |
|
| nsv821643 | Chr.5:69272838 - 71076658 on Build GRCh38 | Loss |
|
| nsv428117 | Chr.5:69347454 - 71500057 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map