Assay Details
Target Gene Details
Entrez Gene ID: | 950 |
Gene Name: | scavenger receptor class B member 2 |
Gene Aliases: |
AMRF, CD36L2, EPM4, HLGP85, LGP85, LIMP-2, LIMPII, SR-BII |
Location: |
Chr.4:76158737-76213899 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCARB2 | NM_001204255.1 | NP_001191184.1 | ||
| NM_005506.3 | NP_005497.1 | |||
| AK022432.1 | 1 | 1087 | ||
| AK296519.1 | ||||
| AK313016.1 | ||||
| BC021892.1 | AAH21892.1 | |||
| BT006939.1 | AAP35585.1 | |||
| D12676.1 | BAA02177.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv995458 | Chr.4:76172611 - 76181025 on Build GRCh38 | Deletion |
|
| esv3601046 | Chr.4:76172675 - 76176770 on Build GRCh38 | Loss |
|
| nsv470047 | Chr.4:76112437 - 76175624 on Build GRCh38 | Gain |
|
| nsv1000178 | Chr.4:76131893 - 76191927 on Build GRCh38 | Gain |
|
| nsv818244 | Chr.4:76173678 - 76175624 on Build GRCh38 | Loss |
|
| nsv461558 | Chr.4:76112572 - 76194400 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map