Assay Details
Target Gene Details
Entrez Gene ID: | 124538 |
Gene Name: | olfactory receptor family 4 subfamily D member 2 |
Gene Aliases: |
BC2009, OR17-24 |
Location: |
Chr.17:58169656-58170579 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR4D2 | NM_001004707.3 | 1 | 647 | NP_001004707.1 |
| AF303373.1 | 1 | 902 | AAL26785.1 |
More Information
Set Membership: |
|

Genomic Map