Assay Details
Target Gene Details
Entrez Gene ID: | 1136 |
Gene Name: | cholinergic receptor nicotinic alpha 3 subunit |
Gene Aliases: |
LNCR2, NACHRA3, PAOD2 |
Location: |
Chr.15:78593052-78621295 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRNA3 | NM_000743.4 | 5 | 1333 | NP_000734.2 |
| NM_001166694.1 | 5 | 1333 | NP_001160166.1 | |
| NR_046313.1 | 5 | 1333 | ||
| XM_006720382.2 | 5 | 1275 | XP_006720445.1 | |
| AF385584.1 | 5 | 816 | AAK68110.1 | |
| BC000513.2 | 5 | 1018 | AAH00513.1 | |
| BC001642.2 | 5 | 1018 | AAH01642.1 | |
| BC002996.1 | 5 | 1018 | AAH02996.1 | |
| BC006114.1 | 5 | 1333 | AAH06114.1 | |
| BC098443.1 | 5 | 1214 | AAH98443.1 | |
| BT006646.1 | 5 | 832 | AAP35292.1 | |
| BT006897.1 | 5 | 832 | AAP35543.1 | |
| M37981.1 | 5 | 895 | AAA59942.1 | |
| U62432.1 | 5 | 1018 | AAB40110.1 | |
| X53559.1 | 4 | 739 | CAA37625.1 | |
| Y08418.1 | 5 | 826 | CAA69695.1 |
More Information
Set Membership: |
|

Genomic Map