Assay Details
Target Gene Details
Entrez Gene ID: | 5978 |
Gene Name: | RE1 silencing transcription factor |
Gene Aliases: |
NRSF, WT6, XBR |
Location: |
Chr.4:56907876-56935847 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REST | NM_001193508.1 | NP_001180437.1 | ||
| NM_005612.4 | NP_005603.3 | |||
| XM_011534401.2 | XP_011532703.1 | |||
| XM_017008527.1 | XP_016864016.1 | |||
| AB209750.1 | BAD92987.1 | |||
| AF228045.1 | AAF61710.1 | |||
| BC017822.1 | ||||
| BC038985.1 | AAH38985.1 | |||
| BC132859.1 | ||||
| BC136491.1 | ||||
| JX896957.1 | ||||
| JX896958.1 | ||||
| JX896959.1 | ||||
| JX896960.1 | ||||
| JX896961.1 | ||||
| JX896962.1 | ||||
| JX896963.1 | ||||
| JX896964.1 | ||||
| JX896965.1 | ||||
| JX896966.1 | ||||
| JX896967.1 | ||||
| JX896968.1 | ||||
| JX896969.1 | ||||
| JX896970.1 | ||||
| JX896971.1 | ||||
| JX896972.1 | ||||
| JX896973.1 | ||||
| JX896974.1 | ||||
| JX896975.1 | ||||
| JX896976.1 | ||||
| JX896977.1 | ||||
| JX896978.1 | ||||
| JX896979.1 | ||||
| JX896980.1 | ||||
| JX896981.1 | ||||
| JX896982.1 | ||||
| JX896983.1 | ||||
| JX896984.1 | ||||
| JX896985.1 | ||||
| JX896986.1 | ||||
| JX896987.1 | ||||
| JX896988.1 | ||||
| JX896989.1 | ||||
| JX896990.1 | ||||
| JX896991.1 | 2 | 1006 | ||
| JX896992.1 | 2 | 910 | ||
| JX896993.1 | 2 | 913 | ||
| KC117262.1 | ||||
| KC117263.1 | ||||
| KC117264.1 | ||||
| KC117265.1 | ||||
| KC117266.1 | ||||
| U13877.1 | 1 | 887 | AAC50114.1 | |
| U13879.1 | AAC50115.1 | |||
| U22314.1 | AAB17211.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5214n100 | Chr.4:56786612 - 56999637 on Build GRCh38 | Gain |
|
| nsv822562 | Chr.4:56902489 - 56913549 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map