Assay Details
Target Gene Details
Entrez Gene ID: | 51329 |
Gene Name: | ADP ribosylation factor like GTPase 6 interacting protein 4 |
Gene Aliases: |
SFRS20, SR-25, SRp25, SRrp37 |
Location: |
Chr.12:122980060-122982913 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL6IP4 | NM_001002251.2 | NP_001002251.2 | ||
| NM_001002252.2 | NP_001002252.2 | |||
| NM_001278378.1 | NP_001265307.1 | |||
| NM_001278379.1 | NP_001265308.1 | |||
| NM_001278380.1 | NP_001265309.1 | |||
| NM_016638.3 | NP_057722.3 | |||
| NM_018694.3 | NP_061164.3 | |||
| NR_103512.1 | ||||
| AB035384.1 | BAA94744.1 | |||
| AF267748.1 | AAF76892.1 | |||
| AK027285.1 | ||||
| AK315759.1 | ||||
| BC001958.1 | AAH01958.1 | |||
| BC015569.2 | AAH15569.1 | |||
| BC015909.1 | AAH15909.2 | |||
| BC094839.1 | AAH94839.1 | |||
| BQ643520.1 | ||||
| CD672894.1 | ||||
| DQ099385.1 | AAZ13761.1 | |||
| EU624490.1 | ||||
| EU624491.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv914 | Chr.12:122977362 - 123008698 on Build GRCh38 | Insertion |
|
| dgv22n68 | Chr.12:122911380 - 123104743 on Build GRCh38 | Loss |
|
| nsv560492 | Chr.12:122943026 - 123090291 on Build GRCh38 | Loss |
|
| nsv517200 | Chr.12:122827504 - 123169951 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map