Assay Details
Target Gene Details
Entrez Gene ID: | 26048 |
Gene Name: | zinc finger protein 500 |
Gene Aliases: |
ZKSCAN18, ZSCAN50 |
Location: |
Chr.16:4745961-4767219 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF500 | NM_001303450.1 | NP_001290379.1 | ||
| NM_021646.3 | 6 | 1586 | NP_067678.1 | |
| XM_005255243.3 | 5 | 1552 | XP_005255300.1 | |
| XM_011522453.2 | XP_011520755.1 | |||
| XM_011522454.2 | 5 | 1341 | XP_011520756.1 | |
| XM_017023121.1 | 5 | 1079 | XP_016878610.1 | |
| AB011129.1 | 5 | 1326 | ||
| AK291792.1 | 6 | 1530 | ||
| AK297993.1 | ||||
| BC004125.2 | 1 | 527 | AAH04125.2 | |
| BC121013.1 | 5 | 1312 | ||
| BC121014.1 | 5 | 1269 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
|
| nsv571302 | Chr.16:4708464 - 4762815 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map