Assay Details
Target Gene Details
Entrez Gene ID: | 4735 |
Gene Name: | septin 2 |
Gene Aliases: |
DIFF6, NEDD-5, NEDD5, Pnutl3, hNedd5 |
Location: |
Chr.2:241315187-241354027 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3594827 | Chr.2:241325760 - 241335933 on Build GRCh38 | Gain |
|
| esv3594826 | Chr.2:241322580 - 241339193 on Build GRCh38 | Gain |
|
| esv2445970 | Chr.2:241325547 - 241326920 on Build GRCh38 | Deletion |
|
| esv2721993 | Chr.2:241326089 - 241326797 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map