Assay Details
Target Gene Details
Entrez Gene ID: | 3851 |
Gene Name: | keratin 4 |
Gene Aliases: |
CK-4, CK4, CYK4, K4, WSN1 |
Location: |
Chr.12:52806543-52814116 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT4 | NM_002272.3 | NP_002263.3 | ||
| AK056254.1 | ||||
| AK098110.1 | ||||
| AK296582.1 | ||||
| AK299394.1 | ||||
| AK299400.1 | ||||
| AK299451.1 | ||||
| AK307941.1 | ||||
| BC003630.2 | AAH03630.2 | |||
| X07695.1 | CAA30534.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832416 | Chr.12:52779656 - 52971582 on Build GRCh38 | Loss |
|
| nsv470293 | Chr.12:52802303 - 52823205 on Build GRCh38 | Loss |
|
| esv3629524 | Chr.12:52688070 - 52835680 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map