Assay Details
Target Gene Details
Entrez Gene ID: | 8462 |
Gene Name: | Kruppel like factor 11 |
Gene Aliases: |
FKLF, FKLF1, MODY7, TIEG2, Tieg3 |
Location: |
Chr.2:10043555-10054836 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLF11 | NM_001177716.1 | 2 | 239 | NP_001171187.1 |
| NM_001177718.1 | 2 | 387 | NP_001171189.1 | |
| NM_003597.4 | 2 | 384 | NP_003588.1 | |
| AF028008.1 | 2 | 239 | AAC61880.1 | |
| AF272830.1 | 2 | 358 | AAF75793.1 | |
| AJ275989.1 | 2 | 222 | CAC06699.1 | |
| AK289816.1 | 2 | 313 | ||
| AK302880.1 | 2 | 352 | ||
| AK315345.1 | 2 | 364 | ||
| AK316439.1 | 2 | 239 | ||
| BC063286.1 | 2 | 384 | AAH63286.1 | |
| BC069383.1 | 2 | 272 | AAH69383.1 | |
| BC074922.2 | 2 | 254 | AAH74922.1 | |
| BG701929.1 | 2 | 394 | ||
| CB990375.1 | 2 | 411 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv828308 | Chr.2:10035398 - 10060201 on Build GRCh38 | Gain |
|
| nsv508826 | Chr.2:10002595 - 10050858 on Build GRCh38 | Insertion |
|
| nsv828297 | Chr.2:9969170 - 10096205 on Build GRCh38 | Loss |
|
| dgv3715n100 | Chr.2:10014833 - 10135998 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map