Assay Details
Target Gene Details
Entrez Gene ID: | 9648 |
Gene Name: | GRIP and coiled-coil domain containing 2 |
Gene Aliases: |
GCC185, RANBP2L4, REN53 |
Location: |
Chr.2:108448561-108509398 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCC2 | NM_181453.3 | NP_852118.1 | ||
| NR_028063.1 | ||||
| XM_006712870.3 | XP_006712933.1 | |||
| XM_006712871.1 | XP_006712934.1 | |||
| XM_006712872.3 | XP_006712935.1 | |||
| XM_011512213.2 | XP_011510515.1 | |||
| AB002334.1 | BAA20794.2 | |||
| AF432211.1 | AAL99918.1 | |||
| AK090813.1 | ||||
| AK308707.1 | ||||
| BC020645.1 | ||||
| BC037774.2 | AAH37774.1 | |||
| BC146789.1 | ||||
| DA865539.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759081 | Chr.2:107833421 - 108606881 on Build GRCh38 | Gain |
|
| nsv834328 | Chr.2:108422875 - 108613866 on Build GRCh38 | Gain |
|
| esv2720498 | Chr.2:107831020 - 108722824 on Build GRCh38 | Deletion |
|
| esv3591913 | Chr.2:107888082 - 108504967 on Build GRCh38 | Gain |
|
| dgv724e199 | Chr.2:107883507 - 108515482 on Build GRCh38 | Deletion |
|
| nsv469774 | Chr.2:108316735 - 108461238 on Build GRCh38 | Gain |
|
| dgv4035n100 | Chr.2:107924531 - 108493578 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73954346] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map