Assay Details
Target Gene Details
Entrez Gene ID: | 9821 |
Gene Name: | RB1 inducible coiled-coil 1 |
Gene Aliases: |
ATG17, CC1, FIP200, PPP1R131 |
Location: |
Chr.8:52622458-52714466 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RB1CC1 | NM_001083617.1 | 24 | 5686 | NP_001077086.1 |
| NM_014781.4 | 24 | 5695 | NP_055596.3 | |
| XM_011517643.2 | 25 | 5884 | XP_011515945.1 | |
| XM_011517644.2 | 24 | 5563 | XP_011515946.1 | |
| XM_011517645.1 | 24 | 5554 | XP_011515947.1 | |
| XM_011517646.2 | 23 | 5534 | XP_011515948.1 | |
| XM_017014103.1 | 24 | 5716 | XP_016869592.1 | |
| XM_017014104.1 | 25 | 5936 | XP_016869593.1 | |
| XM_017014105.1 | 24 | 5707 | XP_016869594.1 | |
| XM_017014106.1 | 24 | 5584 | XP_016869595.1 | |
| XM_017014107.1 | 25 | 5880 | XP_016869596.1 | |
| XM_017014108.1 | 24 | 5575 | XP_016869597.1 | |
| XM_017014109.1 | 23 | 5568 | XP_016869598.1 | |
| XM_017014110.1 | 23 | 5654 | XP_016869599.1 | |
| XM_017014111.1 | 23 | 5633 | XP_016869600.1 | |
| XM_017014112.1 | 23 | 5624 | XP_016869601.1 | |
| AB059622.1 | 24 | 5661 | BAB69690.1 | |
| AK294579.1 | ||||
| BC017556.1 | 23 | 5259 | AAH17556.1 | |
| D86958.1 | 24 | 5643 | ||
| Z35085.1 | 10 | 2033 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv611355 | Chr.8:52490251 - 52776552 on Build GRCh38 | Gain |
|
| dgv7218n100 | Chr.8:52389530 - 53007050 on Build GRCh38 | Gain |
|
| esv3891395 | Chr.8:52330215 - 52674146 on Build GRCh38 | Gain |
|
| esv3576572 | Chr.8:52429401 - 52997384 on Build GRCh38 | Gain |
|
| esv3891396 | Chr.8:52581711 - 52665028 on Build GRCh38 | Gain |
|
| esv3617214 | Chr.8:52513334 - 52648974 on Build GRCh38 | Gain |
|
| esv2764065 | Chr.8:52452089 - 52975372 on Build GRCh38 | Gain |
|
| esv2761192 | Chr.8:52365749 - 52700691 on Build GRCh38 | Gain |
|
| dgv7220n100 | Chr.8:52580301 - 52810024 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map