Assay Details
Target Gene Details
Entrez Gene ID: | 4778 |
Gene Name: | nuclear factor, erythroid 2 |
Gene Aliases: |
NF-E2, p45 |
Location: |
Chr.12:54292107-54301037 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFE2 | NM_001136023.2 | 3 | 482 | NP_001129495.1 |
| NM_001261461.1 | 4 | 612 | NP_001248390.1 | |
| NM_006163.2 | 3 | 481 | NP_006154.1 | |
| XM_005268906.4 | 3 | 332 | XP_005268963.1 | |
| AK290555.1 | 2 | 699 | ||
| AK290564.1 | 2 | 699 | ||
| AK313207.1 | 4 | 590 | ||
| BC005044.1 | 3 | 452 | AAH05044.1 | |
| BT007288.1 | 2 | 189 | AAP35952.1 | |
| BX406317.2 | 3 | 481 | ||
| CR450284.1 | 2 | 189 | CAG29280.1 | |
| L13974.1 | 3 | 445 | AAA35612.1 | |
| L24122.1 | 3 | 309 | AAA16118.1 | |
| S77763.1 | 3 | 462 | AAB34115.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1049160 | Chr.12:54254119 - 54362754 on Build GRCh38 | Gain |
|
| nsv832420 | Chr.12:54137272 - 54296797 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map