Assay Details
Target Gene Details
Entrez Gene ID: | 23406 |
Gene Name: | coactosin like F-actin binding protein 1 |
Gene Aliases: |
CLP |
Location: |
Chr.16:84565596-84618096 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COTL1 | NM_021149.3 | 4 | 1017 | NP_066972.1 |
| AK026033.1 | 3 | 742 | ||
| AK093712.1 | 1 | 1727 | ||
| AK127352.1 | 2 | 4492 | ||
| BC010039.2 | 4 | 984 | AAH10039.1 | |
| BC010884.1 | 4 | 933 | AAH10884.1 | |
| BC016702.1 | 4 | 1002 | AAH16702.1 | |
| BC042970.1 | 4 | 1004 | AAH42970.1 | |
| BC053682.1 | 4 | 980 | AAH53682.1 | |
| L54057.1 | 4 | 984 | AAA88022.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833311 | Chr.16:84489192 - 84663857 on Build GRCh38 | Loss |
|
| dgv5262n54 | Chr.16:84381903 - 84583279 on Build GRCh38 | Loss |
|
| nsv573425 | Chr.16:84298994 - 84575953 on Build GRCh38 | Loss |
|
| nsv1064308 | Chr.16:84461124 - 84592674 on Build GRCh38 | Loss |
|
| nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map