Assay Details
Target Gene Details
Entrez Gene ID: | 4144 |
Gene Name: | methionine adenosyltransferase 2A |
Gene Aliases: |
MATA2, MATII, SAMS2 |
Location: |
Chr.2:85538978-85545280 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAT2A | NM_005911.5 | NP_005902.1 | ||
| AK291126.1 | ||||
| AK293841.1 | ||||
| AK294208.1 | ||||
| AK297758.1 | ||||
| AK297797.1 | ||||
| AK303877.1 | ||||
| AK308758.1 | ||||
| AK309338.1 | ||||
| AK316411.1 | ||||
| BC001686.2 | AAH01686.1 | |||
| BC001854.2 | AAH01854.1 | |||
| DQ083239.1 | AAY85355.1 | |||
| X68836.1 | CAA48726.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2811 | Chr.2:85535833 - 85563891 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map