Assay Details
Target Gene Details
Entrez Gene ID: | 27348 |
Gene Name: | torsin family 1 member B |
Gene Aliases: |
DQ1 |
Location: |
Chr.9:129803143-129811424 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOR1B | NM_001317893.1 | 4 | 1482 | NP_001304822.1 |
| NM_001317894.1 | 3 | 1306 | NP_001304823.1 | |
| NM_014506.2 | 5 | 2495 | NP_055321.1 | |
| NR_133948.1 | 5 | 2636 | ||
| NR_134027.1 | 3 | 2053 | ||
| NR_134113.1 | 5 | 1751 | ||
| XM_005251927.3 | 5 | 1605 | XP_005251984.1 | |
| AF007872.1 | 4 | 2216 | AAC51733.1 | |
| AF317129.1 | 5 | 2485 | AAG50271.1 | |
| BC006480.1 | 1 | 1308 | AAH06480.1 | |
| BC015578.2 | 5 | 2440 | AAH15578.1 | |
| BU633792.1 | 1 | 311 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046477 | Chr.9:129608081 - 129997996 on Build GRCh38 | Gain |
|
| esv3621839 | Chr.9:129701704 - 129885823 on Build GRCh38 | Loss |
|
| nsv6729 | Chr.9:129789659 - 129814916 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77357177] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map