Assay Details
Target Gene Details
Entrez Gene ID: | 79722 |
Gene Name: | ankyrin repeat domain 55 |
Gene Aliases: |
- |
Location: |
Chr.5:56099680-56233359 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD55 | NM_024669.2 | 10 | 1508 | NP_078945.2 |
| XM_011543646.1 | 4 | 822 | XP_011541948.1 | |
| XM_017009852.1 | 10 | 1420 | XP_016865341.1 | |
| XM_017009853.1 | 9 | 1392 | XP_016865342.1 | |
| XM_017009854.1 | 7 | 2742 | XP_016865343.1 | |
| AK021857.1 | 2 | 561 | BAB13916.1 | |
| AK123389.1 | 10 | 1508 | ||
| BC106914.2 | 2 | 728 | AAI06915.1 | |
| BC106915.2 | 2 | 728 | AAI06916.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv329245 | Chr.5:56110251 - 56112504 on Build GRCh38 | Deletion |
|
| nsv1024291 | Chr.5:56070736 - 56111841 on Build GRCh38 | Gain |
|

Genomic Map