Assay Details
Target Gene Details
Entrez Gene ID: | 170685 |
Gene Name: | nudix hydrolase 10 |
Gene Aliases: |
APS2, DIPP3-alpha, DIPP3a |
Location: |
Chr.X:51332231-51337525 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT10 | NM_001304963.1 | 2 | 912 | NP_001291892.1 |
| NM_153183.3 | 3 | 975 | NP_694853.1 | |
| AF469196.1 | 3 | 977 | AAM64113.1 | |
| AK291952.1 | 2 | 892 | ||
| BC049383.1 | 2 | 833 | AAH49383.1 | |
| BC050700.1 | 2 | 891 | AAH50700.1 | |
| BX449265.2 | 2 | 912 | ||
| CN399689.1 | 2 | 581 |
More Information
Set Membership: |
|

Genomic Map