Assay Details
Target Gene Details
Entrez Gene ID: | 10512 |
Gene Name: | semaphorin 3C |
Gene Aliases: |
SEMAE, SemE |
Location: |
Chr.7:80742538-80919351 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEMA3C | NM_006379.3 | 18 | 3170 | NP_006370.1 |
| XM_005250113.2 | 19 | 3091 | XP_005250170.1 | |
| AB000220.1 | 18 | 3170 | BAA32398.1 | |
| AF086506.1 | 1 | 501 | ||
| BC030690.1 | 18 | 2973 | AAH30690.1 | |
| BF983184.1 | ||||
| BM473251.1 | 1 | 153 | ||
| CN334967.1 |
More Information
Set Membership: |
|

Genomic Map