Assay Details
Target Gene Details
Entrez Gene ID: | 889 |
Gene Name: | KRIT1, ankyrin repeat containing |
Gene Aliases: |
CAM, CCM1 |
Location: |
Chr.7:92198969-92246128 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRIT1 | NM_001013406.1 | 17 | 3689 | NP_001013424.1 |
| NM_004912.3 | 19 | 4328 | NP_004903.2 | |
| NM_194454.1 | 19 | 3981 | NP_919436.1 | |
| NM_194455.1 | 19 | 4455 | NP_919437.1 | |
| NM_194456.1 | 20 | 4220 | NP_919438.1 | |
| XM_005250667.2 | 18 | 3739 | XP_005250724.1 | |
| XM_011516653.2 | 18 | 3816 | XP_011514955.1 | |
| XM_017012755.1 | 18 | 3706 | XP_016868244.1 | |
| AF388384.1 | 19 | 4455 | AAM19465.1 | |
| BX648659.1 | 13 | 3960 |
More Information
Set Membership: |
|

Genomic Map