Assay Details
Target Gene Details
Entrez Gene ID: | 9478 |
Gene Name: | calcium binding protein 1 |
Gene Aliases: |
CALBRAIN, HCALB_BR |
Location: |
Chr.12:120640552-120680869 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CABP1 | NM_001033677.1 | NP_001028849.1 | ||
| NM_004276.4 | NP_004267.2 | |||
| NM_031205.3 | NP_112482.1 | |||
| XM_017020235.1 | XP_016875724.1 | |||
| XM_017020236.1 | XP_016875725.1 | |||
| XM_017020237.1 | XP_016875726.1 | |||
| XM_017020238.1 | XP_016875727.1 | |||
| XM_017020239.1 | XP_016875728.1 | |||
| AF169148.1 | AAF25782.1 | |||
| AF169149.1 | AAF25783.1 | |||
| AL533429.3 | ||||
| BC030201.1 | ||||
| BM716422.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045923 | Chr.12:120644400 - 120708416 on Build GRCh38 | Gain |
|
| nsv521928 | Chr.12:120603925 - 121356945 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map