Assay Details
Target Gene Details
Entrez Gene ID: | 684 |
Gene Name: | bone marrow stromal cell antigen 2 |
Gene Aliases: |
CD317, TETHERIN |
Location: |
Chr.19:17402939-17405648 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BST2 | NM_004335.3 | 5 | 983 | NP_004326.1 |
| AI077344.1 | 1 | 56 | ||
| AK223124.1 | BAD96844.1 | |||
| AK291099.1 | 5 | 928 | ||
| AK303593.1 | ||||
| AY320400.1 | ||||
| AY320402.1 | AAP73804.1 | |||
| BC033873.1 | AAH33873.1 | |||
| CB047887.1 | 1 | 71 | ||
| D28137.1 | 5 | 911 | BAA05679.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833768 | Chr.19:17338840 - 17526738 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map