Assay Details
Target Gene Details
Entrez Gene ID: | 389015 |
Gene Name: | solute carrier family 9 member A4 |
Gene Aliases: |
NHE4 |
Location: |
Chr.2:102473282-102533974 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC9A4 | NM_001011552.3 | 1 | 571 | NP_001011552.2 |
| XM_011511158.1 | 1 | 592 | XP_011509460.1 | |
| CR627411.1 | 1 | 572 | CAH10500.1 | |
| DA442134.1 |
More Information
Set Membership: |
|

Genomic Map