Assay Details
Target Gene Details
Entrez Gene ID: | 2635 |
Gene Name: | guanylate binding protein 3 |
Gene Aliases: |
- |
Location: |
Chr.1:89006677-89022880 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GBP3 | NM_001319179.1 | NP_001306108.1 | ||
| NM_001319180.1 | NP_001306109.1 | |||
| NM_001319181.1 | NP_001306110.1 | |||
| NM_018284.2 | NP_060754.2 | |||
| NR_135007.1 | ||||
| XM_005270743.2 | XP_005270800.1 | |||
| XM_006710574.2 | XP_006710637.1 | |||
| XM_011541233.1 | XP_011539535.1 | |||
| XM_011541234.1 | XP_011539536.1 | |||
| XM_017001013.1 | XP_016856502.1 | |||
| XM_017001014.1 | XP_016856503.1 | |||
| AK001823.1 | ||||
| AK309096.1 | ||||
| AL136680.1 | CAB66615.1 | |||
| BC017992.1 | ||||
| BC140837.1 | ||||
| BP281050.1 | ||||
| CR936755.1 | ||||
| CR936758.1 | ||||
| JF927951.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758948 | Chr.1:88946385 - 89095040 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map