Assay Details
Target Gene Details
Entrez Gene ID: | 7111 |
Gene Name: | tropomodulin 1 |
Gene Aliases: |
D9S57E, ETMOD, TMOD |
Location: |
Chr.9:97501180-97601743 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMOD1 | NM_001166116.1 | 10 | 1738 | NP_001159588.1 |
| NM_003275.3 | 10 | 1815 | NP_003266.1 | |
| AF131836.1 | 1 | 1502 | ||
| AK095748.1 | 7 | 1482 | ||
| AK096156.1 | 1 | 1844 | ||
| BC002660.2 | AAH02660.1 | |||
| M77016.1 | 9 | 1636 | AAA61224.1 |
Target Gene Details
Entrez Gene ID: | 158427 |
Gene Name: | thiosulfate sulfurtransferase like domain containing 2 |
Gene Aliases: |
C9orf97 |
Location: |
Chr.9:97600080-97633680 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSTD2 | NM_139246.4 | 10 | 4288 | NP_640339.4 |
| XM_011518312.2 | 9 | 3751 | XP_011516614.1 | |
| XM_017014342.1 | 9 | 3813 | XP_016869831.1 | |
| AK056453.1 | 8 | 3415 | BAB71188.1 | |
| AK056752.1 | 3 | 2849 | BAB71271.1 | |
| AK094043.1 | 11 | 2404 | ||
| AK128615.1 | 11 | 3954 | BAC87530.1 | |
| BC022958.1 | 1 | 2936 | AAH22958.1 | |
| BC035604.1 | 10 | 3935 | AAH35604.1 | |
| BU625565.1 | 1 | 158 | ||
| BX538308.1 | 10 | 3929 | ||
| BX648493.1 | 10 | 3927 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1494e214 | Chr.9:97595020 - 97658603 on Build GRCh38 | Gain |
|
| esv3621190 | Chr.9:97570964 - 97628055 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map