Assay Details
Target Gene Details
Entrez Gene ID: | 84272 |
Gene Name: | Yip1 domain family member 4 |
Gene Aliases: |
FinGER4, Nbla11189 |
Location: |
Chr.2:32277889-32307929 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YIPF4 | NM_032312.3 | NP_115688.1 | ||
| XM_005264599.3 | XP_005264656.1 | |||
| XM_011533125.2 | XP_011531427.1 | |||
| AB072898.1 | BAE45710.1 | |||
| AK098486.1 | BAC05315.1 | |||
| BC004875.2 | AAH04875.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422335 | Chr.2:32187261 - 32975514 on Build GRCh38 | Duplication |
|
| dgv3748n100 | Chr.2:32245383 - 32318300 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map