Assay Details
Target Gene Details
Entrez Gene ID: | 222643 |
Gene Name: | unc-5 family C-terminal like |
Gene Aliases: |
MUXA, ZUD |
Location: |
Chr.6:41026901-41039199 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UNC5CL | NM_173561.2 | NP_775832.2 | ||
| AY510109.1 | AAS80165.1 | |||
| BC035284.1 | AAH35284.1 | |||
| BM312066.1 | ||||
| JF681947.1 |
More Information
Set Membership: |
|

Genomic Map