Assay Details
Target Gene Details
Entrez Gene ID: | 5588 |
Gene Name: | protein kinase C theta |
Gene Aliases: |
PRKCT, nPKC-theta |
Location: |
Chr.10:6393038-6580646 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKCQ | NM_001242413.2 | 17 | 2927 | NP_001229342.1 |
| NM_001282644.1 | 18 | 3114 | NP_001269573.1 | |
| NM_001282645.1 | 17 | 3055 | NP_001269574.1 | |
| NM_001323265.1 | 18 | 3171 | NP_001310194.1 | |
| NM_001323266.1 | 17 | 3053 | NP_001310195.1 | |
| NM_001323267.1 | NP_001310196.1 | |||
| NM_006257.4 | 18 | 3116 | NP_006248.1 | |
| XM_005252497.4 | XP_005252554.1 | |||
| AA234440.1 | 1 | 168 | ||
| AI570478.1 | 1 | 183 | ||
| BF592799.1 | ||||
| BX647657.1 | 17 | 3201 | ||
| R40214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053177 | Chr.10:6411345 - 6431152 on Build GRCh38 | Loss |
|
| nsv1161925 | Chr.10:6417656 - 6429854 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map