Assay Details
Target Gene Details
Entrez Gene ID: | 1558 |
Gene Name: | cytochrome P450 family 2 subfamily C member 8 |
Gene Aliases: |
CPC8, CYPIIC8, MP-12/MP-20 |
Location: |
Chr.10:95036772-95069512 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP2C8 | NM_000770.3 | NP_000761.3 | ||
| NM_001198853.1 | NP_001185782.1 | |||
| NM_001198854.1 | NP_001185783.1 | |||
| NM_001198855.1 | NP_001185784.1 | |||
| AK292753.1 | ||||
| AK293327.1 | ||||
| AK293328.1 | ||||
| AK298401.1 | ||||
| AK303813.1 | ||||
| AK315823.1 | ||||
| BC020596.1 | AAH20596.1 | |||
| CD014119.1 | ||||
| M17397.1 | AAA35739.1 | |||
| M17398.1 | AAA35740.1 | |||
| M21941.1 | AAA52160.1 | |||
| M21942.1 | AAA52161.1 | |||
| Y00498.1 | CAA68550.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551964 | Chr.10:95045614 - 95102529 on Build GRCh38 | Loss |
|
| esv3624264 | Chr.10:94923968 - 95099386 on Build GRCh38 | Gain |
|
| nsv1035409 | Chr.10:94735420 - 95072300 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map