Assay Details
Target Gene Details
Entrez Gene ID: | 5515 |
Gene Name: | protein phosphatase 2 catalytic subunit alpha |
Gene Aliases: |
PP2Ac, PP2CA, PP2Calpha, RP-C |
Location: |
Chr.5:134196457-134226259 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP2CA | NM_002715.2 | 7 | 1558 | NP_002706.1 |
| AK057424.1 | 7 | 1290 | ||
| AK097599.1 | 7 | 1370 | ||
| BC000400.2 | 7 | 1359 | AAH00400.1 | |
| BC002657.2 | 7 | 1359 | AAH02657.1 | |
| BC019275.1 | 7 | 1359 | AAH19275.1 | |
| BC031696.1 | 7 | 1359 | AAH31696.1 | |
| J03804.1 | 7 | 1342 | AAB38019.1 | |
| X12646.1 | 7 | 1243 | CAA31176.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv599732 | Chr.5:134099223 - 134230401 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map