Assay Details
Target Gene Details
Entrez Gene ID: | 126306 |
Gene Name: | junctional sarcoplasmic reticulum protein 1 |
Gene Aliases: |
JP-45, JP45 |
Location: |
Chr.19:2252251-2260813 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JSRP1 | NM_144616.3 | NP_653217.1 | ||
| XM_017026285.1 | XP_016881774.1 | |||
| XM_017026286.1 | XP_016881775.1 | |||
| XM_017026287.1 | XP_016881776.1 | |||
| AK056978.1 | BAB71330.1 | |||
| BC021201.2 | AAH21201.1 | |||
| DA897543.1 | ||||
| DA898560.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv578382 | Chr.19:2194897 - 2260362 on Build GRCh38 | Loss |
|
| dgv6218n54 | Chr.19:2221793 - 2336009 on Build GRCh38 | Loss |
|
| nsv578383 | Chr.19:2214058 - 2285197 on Build GRCh38 | Loss |
|
| nsv833707 | Chr.19:2187092 - 2372223 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map