Assay Details
Target Gene Details
Entrez Gene ID: | 79739 |
Gene Name: | tubulin tyrosine ligase like 7 |
Gene Aliases: |
- |
Location: |
Chr.1:83865021-83999340 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTLL7 | NM_024686.4 | 21 | 3083 | NP_078962.4 |
| XM_005271208.2 | 20 | 2894 | XP_005271265.1 | |
| XM_017002347.1 | 22 | 3103 | XP_016857836.1 | |
| XM_017002349.1 | 20 | 2858 | XP_016857838.1 | |
| AK026686.1 | 14 | 1849 | BAB15526.1 | |
| AK126792.1 | 21 | 2980 | BAC86695.1 | |
| AK128604.1 | 18 | 3058 | BAC87524.1 | |
| AK225574.1 | 14 | 1849 | ||
| AY170843.1 | 20 | 2917 | AAO37763.1 | |
| BC028350.2 | 16 | 2246 | AAH28350.2 |
More Information
Set Membership: |
|

Genomic Map