Assay Details
Target Gene Details
Entrez Gene ID: | 374786 |
Gene Name: | EF-hand calcium binding domain 5 |
Gene Aliases: |
- |
Location: |
Chr.17:29929856-30108452 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFCAB5 | NM_001145053.1 | NP_001138525.1 | ||
| NM_198529.3 | NP_940931.2 | |||
| NR_026738.1 | ||||
| XM_011524759.1 | XP_011523061.1 | |||
| XM_011524763.2 | XP_011523065.1 | |||
| XM_011524765.1 | XP_011523067.1 | |||
| XM_011524768.2 | XP_011523070.1 | |||
| XM_017024592.1 | XP_016880081.1 | |||
| AK299608.1 | ||||
| AK302745.1 | ||||
| AK303060.1 | ||||
| BC137519.1 | ||||
| BX648689.1 | ||||
| DB064946.1 | ||||
| DB080627.1 |
More Information
Set Membership: |
|

Genomic Map