Assay Details
Target Gene Details
Entrez Gene ID: | 26689 |
Gene Name: | olfactory receptor family 4 subfamily D member 1 |
Gene Aliases: |
OR17-23, OR4D3, OR4D4P, TPCR16 |
Location: |
Chr.17:58155154-58156086 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR4D1 | NM_012374.1 | 1 | 220 | NP_036506.1 |
| BC136589.1 | 1 | 241 | ||
| BC136595.1 | 1 | 241 |
More Information
Set Membership: |
|

Genomic Map