Assay Details
Target Gene Details
Entrez Gene ID: | 3606 |
Gene Name: | interleukin 18 |
Gene Aliases: |
IGIF, IL-18, IL-1g, IL1F4 |
Location: |
Chr.11:112143251-112164117 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL18 | NM_001243211.1 | NP_001230140.1 | ||
| NM_001562.3 | NP_001553.1 | |||
| XM_011542805.1 | XP_011541107.1 | |||
| XM_011542806.2 | XP_011541108.1 | |||
| AF077611.1 | AAC27787.1 | |||
| AF380360.1 | AAK57024.1 | |||
| AK311301.1 | ||||
| AY044641.1 | AAK95950.1 | |||
| AY266351.1 | AAP92112.1 | |||
| BC007007.1 | AAH07007.1 | |||
| BC007461.1 | AAH07461.1 | |||
| CR541973.1 | CAG46771.1 | |||
| CR542001.1 | CAG46798.1 | |||
| D49950.1 | BAA08706.1 | |||
| U90434.1 | AAB50010.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745077 | Chr.11:111949712 - 112188677 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map