Assay Details
Target Gene Details
Entrez Gene ID: | 219858 |
Gene Name: | olfactory receptor family 8 subfamily B member 12 |
Gene Aliases: |
OR11-317 |
Location: |
Chr.11:124542722-124543654 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR8B12 | NM_001005195.1 | 1 | 185 | NP_001005195.1 |
| BC136863.1 | 1 | 210 | ||
| BC136864.1 | 1 | 210 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518199 | Chr.11:124516115 - 124580084 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map