Assay Details
Target Gene Details
Entrez Gene ID: | 7272 |
Gene Name: | TTK protein kinase |
Gene Aliases: |
CT96, ESK, MPH1, MPS1, MPS1L1, PYT |
Location: |
Chr.6:80004147-80043577 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTK | NM_001166691.1 | 6 | 792 | NP_001160163.1 |
| NM_003318.4 | 6 | 780 | NP_003309.2 | |
| XM_011536099.2 | 6 | 754 | XP_011534401.1 | |
| XM_011536100.2 | 6 | 780 | XP_011534402.1 | |
| XM_017011242.1 | 7 | 1309 | XP_016866731.1 | |
| AK095599.1 | 6 | 1252 | ||
| AK292460.1 | 6 | 755 | ||
| AK315696.1 | 6 | 697 | ||
| AX775733.1 | 6 | 752 | CAE11566.1 | |
| BC000633.2 | 6 | 743 | AAH00633.1 | |
| BC032858.2 | 6 | 745 | AAH32858.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5370 | Chr.6:80005634 - 80042023 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map