Assay Details
Target Gene Details
Entrez Gene ID: | 10313 |
Gene Name: | reticulon 3 |
Gene Aliases: |
ASYIP, HAP, NSPL2, NSPLII, RTN3-A1 |
Location: |
Chr.11:63681315-63759891 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RTN3 | NM_001265589.1 | NP_001252518.1 | ||
| NM_001265590.1 | NP_001252519.1 | |||
| NM_001265591.1 | NP_001252520.1 | |||
| NM_006054.3 | NP_006045.1 | |||
| NM_201428.2 | NP_958831.1 | |||
| NM_201429.2 | NP_958832.1 | |||
| NM_201430.2 | NP_958833.1 | |||
| NR_049750.1 | ||||
| NR_049751.1 | ||||
| XM_011544730.2 | 1 | 119 | XP_011543032.1 | |
| XM_011544731.2 | 1 | 164 | XP_011543033.1 | |
| XM_017017091.1 | 1 | 119 | XP_016872580.1 | |
| XM_017017092.1 | 1 | 119 | XP_016872581.1 | |
| AK075412.1 | ||||
| AK295534.1 | ||||
| AK297533.1 | ||||
| AK304100.1 | ||||
| AK307811.1 | ||||
| AK308083.1 | ||||
| AK309766.1 | ||||
| AY427821.2 | AAR02474.1 | |||
| AY750848.1 | AAU81930.1 | |||
| BC011394.1 | AAH11394.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832183 | Chr.11:63649206 - 63797093 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map