Assay Details
Target Gene Details
Entrez Gene ID: | 80070 |
Gene Name: | ADAM metallopeptidase with thrombospondin type 1 motif 20 |
Gene Aliases: |
ADAM-TS20, ADAMTS-20, GON-1 |
Location: |
Chr.12:43352843-43551971 on Build GRCh38 |
Assay Gene Location: | Within Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAMTS20 | NM_025003.3 | 33 | 5063 | NP_079279.3 |
| XM_011538754.2 | 33 | 5116 | XP_011537056.1 | |
| XM_017019979.1 | 26 | 4132 | XP_016875468.1 | |
| AF488804.1 | 33 | 5066 | AAO15766.1 | |
| AJ515153.3 | 33 | 5057 | CAD56159.3 |
More Information
Set Membership: |
|

Genomic Map