Assay Details
Target Gene Details
Entrez Gene ID: | 3624 |
Gene Name: | inhibin beta A subunit |
Gene Aliases: |
EDF, FRP |
Location: |
Chr.7:41685101-41710532 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INHBA | NM_002192.2 | NP_002183.1 | ||
| XM_017012174.1 | XP_016867663.1 | |||
| XM_017012175.1 | XP_016867664.1 | |||
| XM_017012176.1 | XP_016867665.1 | |||
| AU137531.1 | ||||
| BC007858.2 | AAH07858.1 | |||
| D49743.1 | BAA08577.1 |
Target Gene Details
Entrez Gene ID: | 285954 |
Gene Name: | INHBA antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.7:41693916-41779378 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INHBA-AS1 | NR_027118.1 | |||
| NR_027119.1 |
More Information
Set Membership: |
|

Genomic Map