Assay Details
Target Gene Details
Entrez Gene ID: | 5836 |
Gene Name: | phosphorylase, glycogen, liver |
Gene Aliases: |
GSD6 |
Location: |
Chr.14:50905217-50944530 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYGL | NM_001163940.1 | NP_001157412.1 | ||
| NM_002863.4 | NP_002854.3 | |||
| AA523002.1 | ||||
| AF046785.1 | AAC23504.1 | |||
| AF066858.1 | AAC17450.1 | |||
| AK225904.1 | ||||
| AK300580.1 | ||||
| AK309333.1 | ||||
| AK313206.1 | ||||
| BC009895.2 | ||||
| BC082229.1 | ||||
| BC095850.1 | ||||
| BC110791.1 | ||||
| M14636.1 | AAA52577.1 | |||
| M36807.1 | AAA35906.1 | |||
| Y15233.1 | CAA75517.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892542 | Chr.14:50853532 - 50942040 on Build GRCh38 | Loss |
|
| nsv832794 | Chr.14:50739915 - 50959891 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map