Assay Details
Target Gene Details
Entrez Gene ID: | 55454 |
Gene Name: | chondroitin sulfate N-acetylgalactosaminyltransferase 2 |
Gene Aliases: |
CHGN2, ChGn-2, GALNACT-2, GALNACT2, PRO0082, beta4GalNAcT |
Location: |
Chr.10:43138445-43185308 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSGALNACT2 | NM_001319654.1 | NP_001306583.1 | ||
| NM_001319656.1 | NP_001306585.1 | |||
| NM_018590.4 | NP_061060.3 | |||
| NR_135077.1 | ||||
| XM_011539938.1 | XP_011538240.1 | |||
| XM_017016388.1 | XP_016871877.1 | |||
| XM_017016389.1 | XP_016871878.1 | |||
| XM_017016390.1 | XP_016871879.1 | |||
| XM_017016391.1 | XP_016871880.1 | |||
| AB079252.1 | BAC55935.1 | |||
| AB090811.1 | BAC55936.1 | |||
| AK125300.1 | ||||
| BC030268.1 | AAH30268.1 | |||
| BX640967.1 | CAE45982.1 | |||
| CT003455.1 | ||||
| KU178704.1 | ||||
| KU178705.1 | ||||
| KU178706.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951565 | Chr.10:43137753 - 43161752 on Build GRCh38 | Duplication |
|
| nsv1042310 | Chr.10:43127612 - 43193057 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map