Assay Details
Target Gene Details
Entrez Gene ID: | 196951 |
Gene Name: | family with sequence similarity 227 member B |
Gene Aliases: |
C15orf33 |
Location: |
Chr.15:49326997-49620931 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636427 | Chr.15:49359900 - 49397828 on Build GRCh38 | Loss |
|
| nsv833002 | Chr.15:49329203 - 49489602 on Build GRCh38 | Gain |
|
| nsv442375 | Chr.15:49360327 - 49391885 on Build GRCh38 | Loss |
|
| esv3636426 | Chr.15:49354085 - 49408851 on Build GRCh38 | Loss |
|

Genomic Map