Assay Details
Target Gene Details
Entrez Gene ID: | 390882 |
Gene Name: | olfactory receptor family 7 subfamily G member 2 |
Gene Aliases: |
OR19-6, OST260 |
Location: |
Chr.19:9102269-9103306 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR7G2 | NM_001005193.1 | 1 | 400 | NP_001005193.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056401 | Chr.19:9031263 - 9161005 on Build GRCh38 | Gain |
|
| nsv509720 | Chr.19:9072760 - 9188811 on Build GRCh38 | Insertion |
|
| esv2762024 | Chr.19:8567703 - 9197018 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map